1-((4-Methylphenyl)sulfonyl)-4-pyridinecarboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: 1-Tosylpiperidine-4-carboxylic acid

A166053-100MG A166053-10G A166053-5G A166053-1G A166053-250MG
103806-014EA 109 USD
103806-014 103844-270 76720-306 76720-308 77300-806 77320-500 77667-328
1-((4-Methylphenyl)sulfonyl)-4-pyridinecarboxylic acid 95%
1-((4-Methylphenyl)sulphonyl)-4-pyridinecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₇NO₄S
MW: 283.3400 g/mol
Boiling Pt: 477.6 °C (760 mm Hg)
Melting Pt: 167…169 °C
Flash Pt: 242.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00723645
CAS Number: 147636-36-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR