1-Boc-isonipecotic acid methyl ester 98%

Supplier: Ambeed
Warning

Synonyms: Methyl piperidine-4-carboxylate, N-BOC protected, 1-tert-Butyl 4-methyl piperidine-1,4-dicarboxylate

A429558-1000G A429558-100G A429558-10G A429558-500G A429558-25G A429558-5G
103838-562EA 5 USD
103838-562 76725-336 76725-338 76725-340 76725-342 76842-250 76842-252
1-Boc-isonipecotic acid methyl ester 98%
1-Boc-isonipecotic acid methyl ester

Precaución: for Research Use Only

Formula: C₁₂H₂₁NO₄
MW: 243.2994 g/mol
Boiling Pt: 307.4 °C (760 mm Hg)
Flash Pt: 139.7 °C
Storage Temperature: Ambient
MDL Number: MFCD02183584
CAS Number: 124443-68-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR