1-tert-Butyl 4-methyl 5,6-dihydropyridine-1,4(2H)-dicarboxylate 97%

Supplier: Ambeed
Warning

A376718-1G A376718-5G A376718-100MG
103826-590EA 139 USD
103826-590 103837-616 76840-918 76840-920 77168-032
1-tert-Butyl 4-methyl 5,6-dihydropyridine-1,4(2H)-dicarboxylate 97%
1-tert-Butyl 4-methyl 5,6-dihydropyridine-1,4(2H)-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₉NO₄
Storage Temperature: Refrigerator
MDL Number: MFCD23704701
CAS Number: 184368-74-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR