1-tert-Butyl-4-ethyl-3-oxopiperidine-1,4-dicarboxylate 97%

Supplier: Ambeed
Warning

Synonyms: 1-tert-Butyl 4-ethyl 3-oxopiperidine-1,4-dicarboxylate, Ethyl 1-Boc-3-oxopiperidine-4-carboxylate

A116287-100G A116287-10G A116287-25G A116287-1G A116287-5G
103798-708EA 9 USD
103798-708 103824-570 77284-538 77284-540 77284-542 77297-448 77311-386
1-tert-Butyl-4-ethyl-3-oxopiperidine-1,4-dicarboxylate 97%
1-tert-Butyl-4-ethyl-3-oxopiperidine-1,4-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₃H₂₁NO₅
MW: 271.31 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09878815
CAS Number: 71233-25-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR