1-tert-Butyl-3-methylpiperazine-1,3-dicarboxylate 97%

Supplier: Ambeed
Warning

Synonyms: 1-tert-Butyl 3-methyl piperazine-1,3-dicarboxylate

A112145-100G A112145-10G A112145-250MG A112145-25G A112145-1G A112145-5G
103797-960EA 14 USD
103797-960 103823-794 76796-914 76796-916 76796-918 76796-920 77310-600 77310-602
1-tert-Butyl-3-methylpiperazine-1,3-dicarboxylate 97%
1-tert-Butyl-3-methylpiperazine-1,3-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₁H₂₀N₂O₄
MW: 244.29 g/mol
Boiling Pt: 321.3 °C (760 mm Hg)
Flash Pt: 148.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01632465
CAS Number: 129799-08-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR