1-tert-Butyl-3-ethyl-4-oxopiperidine-1,3-dicarboxylate 97%

Supplier: Ambeed
Warning

A149124-100G A149124-10G A149124-250MG A149124-1G A149124-25G A149124-5G A149124-500G
103804-002EA 6 USD
103804-002 103842-126 77150-320 77150-322 77150-324 77299-704 77317-608 77317-610 77685-247
1-tert-Butyl-3-ethyl-4-oxopiperidine-1,3-dicarboxylate 97%
1-tert-Butyl-3-ethyl-4-oxopiperidine-1,3-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₃H₂₁NO₅
Boiling Pt: 364.6 °C (760 mm Hg)
Melting Pt: 50…54 °C
Flash Pt: 174.3 °C
Storage Temperature: Ambient
MDL Number: MFCD01862189
CAS Number: 98977-34-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR