1-tert-Butyl 2-methyl pyrrolidine-1,2-dicarboxylate 97%

Supplier: Ambeed
Warning

A134073-100G A134073-10G A134073-500G A134073-1G A134073-5G
103801-966EA 5 USD
103801-966 76811-392 76811-394 76811-396 77314-678 77314-680
1-tert-Butyl 2-methyl pyrrolidine-1,2-dicarboxylate 97%
1-tert-Butyl 2-methyl pyrrolidine-1,2-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₄
MW: 229.27 g/mol
Boiling Pt: 288.6 °C (760 mm Hg)
Flash Pt: 128.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03762834
CAS Number: 145681-01-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR