Methyl-1-(tert-butoxycarbonyl)-piperidine-2-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 1-Boc-2-piperidinecarboxylate, 1-Boc-2-piperidinecarboxylic acid methyll ester, Methyl 1-(tert-Butoxycarbonyl)-2-piperidinecarboxylate, Piperidine-1,2-dicarboxylic acid 1-tert-Butyl 2-Methyl ester, 1-tert-Butyl 2-Methyl Piperidine-1,2-dicarboxylate, 1-(tert-Butoxycarbonyl)-2-piperidinecarboxylic acid methyll ester

A138634-100G A138634-10G A138634-500G
77010-246EA 63.66 USD
77010-246 77010-248 77010-250
Methyl-1-(tert-butoxycarbonyl)-piperidine-2-carboxylate 97%
Methyl-1-(tert-butoxycarbonyl)-piperidine-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₂₁NO₄
MW: 243.31 g/mol
Boiling Pt: 307.4 °C (760 mm Hg)
Flash Pt: 139.7 °C
Storage Temperature: Ambient
MDL Number: MFCD06795925
CAS Number: 167423-93-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR