1-tert-Butyl 2-methyl 4-oxopiperidine-1,2-dicarboxylate 95%

Supplier: Ambeed
Warning

Synonyms: 1-(tert-Butoxycarbonyl)-2-(methoxycarbonyl)-4-oxopiperidine, Methyl 4-oxopiperidine-2-carboxylate, N-BOC protected

A276890-100MG A276890-250MG A276890-1G A276890-25G A276890-5G
103813-326EA 30 USD
103813-326 103853-262 77032-070 77032-072 77093-128 77093-130 77093-132
1-tert-Butyl 2-methyl 4-oxopiperidine-1,2-dicarboxylate 95%
1-tert-Butyl 2-methyl 4-oxopiperidine-1,2-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₉NO₅
MW: 257.283 g/mol
Boiling Pt: 350 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09842531
CAS Number: 81357-18-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR