1-tert-Butyl 2-ethyl pyrrolidine-1,2-dicarboxylate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Boc-DL-Pro-OEt
A136955-10G
A136955-1G
A136955-25G
A136955-5G
103802-382EA
9
USD
103802-382
76688-838
77315-310
77315-312
77315-314
1-tert-Butyl 2-ethyl pyrrolidine-1,2-dicarboxylate 98%
1-tert-Butyl 2-ethyl pyrrolidine-1,2-dicarboxylate
Precaución: for Research Use Only
Formula:
C₁₂H₂₁NO₄ MW: 243.3 g/mol Boiling Pt: 305.6 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD06656725 CAS Number: 125347-83-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...