(1H-Pyrazol-1-yl)acetic acid 95%

Supplier: Ambeed
Warning

Synonyms: 1-Pyrazoleacetic Acid

A140364-10G A140364-250MG A140364-25G A140364-5G
103802-858EA 23 USD
103802-858 77234-902 77234-904 77299-130 77299-132
(1H-Pyrazol-1-yl)acetic acid 95%
(1H-Pyrazol-1-yl)acetic acid

Precaución: for Research Use Only

Formula: C₅H₆N₂O₂
MW: 126.1133 g/mol
Boiling Pt: 301.1 °C (760 mm Hg)
Melting Pt: 170…174 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00228774
CAS Number: 16034-48-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR