1-Phenyl-1H-pyrazole-4-carbaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 1-Phenylpyrazole-4-carboxaldehyde, 1-Phenyl-1H-pyrazole-4-carboxaldehyde

A475806-5G A475806-100MG A475806-10G A475806-250MG
103828-078EA 103 USD
103828-078 103839-148 76843-076 77169-076 77169-078 77169-080
1-Phenyl-1H-pyrazole-4-carbaldehyde 98%
1-Phenyl-1H-pyrazole-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₈N₂O
MW: 172.18 g/mol
Boiling Pt: 312.7 °C (760 mm Hg)
Melting Pt: 85…88 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02179568
CAS Number: 54605-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR