(±)-1-Phenyl-1,2-ethanediol 97%

Supplier: Ambeed
Warning

Synonyms: (±)-α,β-Dihydroxyethylbenzene, (±)-Phenylethane-1,2-diol, 1-Phenylethane-1,2-diol

A626063-25G A626063-5G A626063-10G
103840-252EA 7 USD
103840-252 76844-412 76844-414 77242-826
(±)-1-Phenyl-1,2-ethanediol 97%
(±)-1-Phenyl-1,2-ethanediol

Precaución: for Research Use Only

Formula: C₆H₅CH(OH)CH₂OH
MW: 138.17 g/mol
Boiling Pt: 272…274 °C (760 mm Hg)
Melting Pt: 61…67 °C
Flash Pt: 145.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00003546
CAS Number: 93-56-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR