(±)-1-Phenyl-2-propyn-1-ol 98%
Supplier: Ambeed
|
Warning
|
A156799-100G
A156799-10G
A156799-25G
A156799-1G
A156799-5G
103804-924EA
6
USD
103804-924
103843-126
77235-696
77235-698
77300-218
77318-926
77318-928
(±)-1-Phenyl-2-propyn-1-ol 98%
(±)-1-Phenyl-2-propyn-1-ol
Precaución: for Research Use Only
Formula:
C₉H₈O Boiling Pt: 231 °C Melting Pt: 22…24 °C Flash Pt: 99 °C Storage Temperature: Ambient |
MDL Number:
MFCD00021860 CAS Number: 4187-87-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...