1-Phenyl-2-propanol 98%

Supplier: Ambeed
Warning

Synonyms: (2-Hydroxypropyl)benzol, (±)-1-Phenyl-2-propanol, (±)-α-Methylphenethyl alcohol, Benzylmethylcarbinol

A158681-10G A158681-1G A158681-25G A158681-5G
103805-156EA 6 USD
103805-156 103843-370 77235-748 77300-348 77300-350 77319-230
1-Phenyl-2-propanol 98%
1-Phenyl-2-propanol

Precaución: for Research Use Only

Formula: C₆H₅CH₂CH(OH)CH₃
MW: 136.19 g/mol
Melting Pt: 135…136 °C at N/A mmHg
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00004540
CAS Number: 698-87-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR