1-Phenyl-1H-pyrazol-3-amine 97%

Supplier: Ambeed
Warning

A235878-5G A235878-100MG A235878-250MG A235878-1G
103811-040EA 48 USD
103811-040 103850-810 77089-828 77238-006 77238-008 77304-020
1-Phenyl-1H-pyrazol-3-amine 97%
1-Phenyl-1H-pyrazol-3-amine

Precaución: for Research Use Only

Formula: C₉H₉N₃
Boiling Pt: 323.2 °C (760 mm Hg)
Flash Pt: 149.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00100520
CAS Number: 1128-56-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR