1-Phenylimidazole 98%

Supplier: Ambeed
Warning

A469615-10G A469615-5G A469615-1G A469615-25G
103828-012EA 9 USD
103828-012 103839-086 76818-502 76842-992 77307-468 77307-470
1-Phenylimidazole 98%
1-Phenylimidazole

Precaución: for Research Use Only

Formula: C₉H₈N₂
MW: 144.18 g/mol
Boiling Pt: 142 °C (760 mm Hg)
Melting Pt: 13 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00041204
CAS Number: 7164-98-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR