1-Phenyl-1H-benzo[d]imidazol-2(3H)-one 98%

Supplier: Ambeed
Warning

A145795-100MG A145795-10G A145795-250MG A145795-5G
103803-592EA 38 USD
103803-592 103841-672 76823-642 76823-644 76823-646 77316-974
1-Phenyl-1H-benzo[d]imidazol-2(3H)-one 98%
1-Phenyl-1H-benzo[d]imidazol-2(3H)-one

Precaución: for Research Use Only

Formula: C₁₃H₁₀N₂O
MW: 210.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01658803
CAS Number: 14813-85-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR