1-Octyl-1H-imidazole 96%

Supplier: Ambeed
Warning

A129813-100G A129813-10G A129813-1G
103801-334EA 6 USD
103801-334 77234-470 77234-472 77298-372
1-Octyl-1H-imidazole 96%
1-Octyl-1H-imidazole

Precaución: for Research Use Only

Formula: C₁₁H₂₀N₂
MW: 180.29 g/mol
Boiling Pt: 302.4 °C (760 mm Hg)
Flash Pt: 136.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00467256
CAS Number: 21252-69-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR