1-N-Cbz-pyrrolidine-3-carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 1-(Benzyloxycarbonyl)pyrrolidine-3-carboxylic acid, 1-N-Cbz-Pyrrolidine-3-carboxylic acid

A131638-10G A131638-250MG A131638-25G A131638-1G
103801-614EA 13 USD
103801-614 77094-986 77094-988 77298-504 77314-120
1-N-Cbz-pyrrolidine-3-carboxylic acid 97%
1-N-Cbz-pyrrolidine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₅NO₄
MW: 249.27 g/mol
Boiling Pt: 432.3 °C (760 mm Hg)
Melting Pt: 78…81 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02179036
CAS Number: 188527-21-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR