1-N-Cbz-4-aminopiperidine 98%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-1-piperidinecarboxylic acid Benzyl ester, 4-Amino-1-benzyloxycarbonylpiperidine, 4-Amino-1-carbobenzoxypiperidine, Benzyl 4-aminopiperidine-1-carboxylate, 4-Amino-1-Cbz-piperidine, 1-N-Cbz-4-Aminopiperidine, Benzyl 4-Amino-1-piperidinecarboxylate

A125525-10G A125525-250MG A125525-25G A125525-1G A125525-5G A125525-100G
103800-690EA 7 USD
103800-690 77149-412 77149-414 77149-416 77312-836 77312-838 77549-484
1-N-Cbz-4-aminopiperidine 98%
1-N-Cbz-4-aminopiperidine

Precaución: for Research Use Only

Formula: C₁₃H₁₈N₂O₂
MW: 234.30 g/mol
Boiling Pt: 367.2 °C (760 mm Hg)
Flash Pt: 175.9 °C
Storage Temperature: Ambient
MDL Number: MFCD05863884
CAS Number: 120278-07-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR