1-N-Boc-pyrrolidin-2-ylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 1-N-Boc-Pyrrolidin-2-ylboronic acid, 2-Borono-1-pyrrolidinecarboxylic acid 1-(1,1-dimethylethyl) ester

A149947-100MG A149947-250MG A149947-25G
103804-108EA 18 USD
103804-108 103842-236 77259-218 77259-220 77746-041
1-N-Boc-pyrrolidin-2-ylboronic acid 97%
1-N-Boc-pyrrolidin-2-ylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₈BNO₄
MW: 215.0545 g/mol
Boiling Pt: 349.2 °C (760 mm Hg)
Storage Temperature: Freezer
MDL Number: MFCD02183523
CAS Number: 149682-75-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR