1-N-Boc-4-(4-bromophenyl)piperidine 98%
Supplier: Ambeed
|
Warning
|
A140200-10G
A140200-250MG
A140200-1G
A140200-25G
A140200-5G
103802-846EA
12
USD
103802-846
76782-900
76782-902
77315-964
77315-966
77315-968
1-N-Boc-4-(4-bromophenyl)piperidine 98%
1-N-Boc-4-(4-bromophenyl)piperidine
Precaución: for Research Use Only
Formula:
C₁₆H₂₂BrNO₂ Boiling Pt: 398.5 °C (760 mm Hg) Flash Pt: 170.887 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD09878774 CAS Number: 769944-78-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...