1-Boc-3-cyanopiperidine 97%

Supplier: Ambeed
Warning

Synonyms: tert-butyl 3-cyanopiperidine-1-carboxylate, 1-Boc-3-cyanopiperidine, 3-Cyano-1-piperidinecarboxylic acid tert-Butyl ester, tert-Butyl 3-Cyano-1-piperidinecarboxylate, 1-Boc-3-piperidinecarbonitrile, 1-tert-Butoxycarbonyl-3-piperidinecarbonitrile, 1-tert-Butoxycarbonyl-3-cyanopiperidine

A129293-100G A129293-10G A129293-25G A129293-1G A129293-5G
103801-258EA 6 USD
103801-258 76797-802 76797-804 77298-334 77313-640 77313-642
1-Boc-3-cyanopiperidine 97%
1-Boc-3-cyanopiperidine

Precaución: for Research Use Only

Formula: C₁₁H₁₈N₂O₂
MW: 210.27 g/mol
Melting Pt: 60…65 °C
Flash Pt: 150.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01861222
CAS Number: 91419-53-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR