1-Naphthoic acid 98%

Supplier: Ambeed
Warning

Synonyms: α-Naphthoic acid

A173386-100G A173386-10G A173386-500G A173386-25G A173386-5G
103845-112EA 5 USD
103845-112 76872-502 76872-504 76872-506 77301-262 77301-264
1-Naphthoic acid 98%
1-Naphthoic acid

Precaución: for Research Use Only

Formula: C₁₁H₈O₂
MW: 172.18 g/mol
Boiling Pt: 300 °C (760 mm Hg)
Melting Pt: 157…160 °C
Storage Temperature: Ambient
MDL Number: MFCD00004007
CAS Number: 86-55-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR