1-Methylpyrrolidine-3-carboxylic acid hydrochloride 95%

Supplier: Ambeed
Warning

A234576-1G A234576-5G A234576-10G A234576-250MG
103810-960EA 50 USD
103810-960 103850-716 77089-706 77089-708 77165-650 77165-652
1-Methylpyrrolidine-3-carboxylic acid hydrochloride 95%
1-Methylpyrrolidine-3-carboxylic acid hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₂ClNO₂
MW: 165.6200 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06738921
CAS Number: 50585-87-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR