1-Methyl-2-pyridone 98%

Supplier: Ambeed
Warning

A151491-100G A151491-10G A151491-250MG A151491-25G A151491-5G
103804-260EA 6 USD
103804-260 103842-412 77235-448 77235-450 77235-452 77299-882 77317-978
1-Methyl-2-pyridone 98%
1-Methyl-2-pyridone

Precaución: for Research Use Only

Formula: C₆H₇NO
MW: 109.13 g/mol
Boiling Pt: 250 °C (760 mm Hg)
Melting Pt: 30…32 °C
Flash Pt: 125.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00006520
CAS Number: 694-85-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR