1-Methylpyridin-1-ium iodide 95%

Supplier: Ambeed
Danger

A203735-250MG A203735-1G
103817-484EA 21 USD
103817-484 103848-144 77164-682 77302-866
1-Methylpyridin-1-ium iodide 95%
1-Methylpyridin-1-ium iodide

Precaución: for Research Use Only

Formula: C₆H₈IN
MW: 221.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00160400
CAS Number: 930-73-4
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR