1-Methylpiperidine-4-carboxylic acid 95%

Supplier: Ambeed
Warning

A177267-1G A177267-25G A177267-5G A177267-100G A177267-10G
103807-222EA 5 USD
103807-222 103845-550 77082-942 77082-944 77082-946 77111-262 77567-610
1-Methylpiperidine-4-carboxylic acid 95%
1-Methylpiperidine-4-carboxylic acid

Precaución: For research use only.

Formula: C₇H₁₃NO₂
Boiling Pt: 246.1 °C (760 mm Hg)
Flash Pt: 102.6 °C
Storage Temperature: Ambient
MDL Number: MFCD06659473
CAS Number: 68947-43-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR