1-Methylpiperazin-2-one 97%
Supplier: Ambeed
|
Warning
|
A245378-1G
A245378-5G
A245378-100MG
A245378-10G
A245378-250MG
A245378-25G
103811-640EA
17
USD
103811-640
103851-476
77090-722
77090-724
77165-860
77165-862
77165-864
77304-360
1-Methylpiperazin-2-one 97%
1-Methylpiperazin-2-one
Precaución: for Research Use Only
Formula:
C₅H₁₀N₂O MW: 114.1457 g/mol Boiling Pt: 242 °C (760 mm Hg) Flash Pt: 100.231 °C Storage Temperature: Ambient |
MDL Number:
MFCD00102572 CAS Number: 59702-07-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...