1-Methylimidazolidin-2-one 97%

Supplier: Ambeed
Warning

Synonyms: 1-Methyl-2-imidazolidinone

A202167-1G A202167-250MG A202167-5G A202167-10G
103817-360EA 24 USD
103817-360 103847-996 77086-094 77237-186 77302-792 77746-261
1-Methylimidazolidin-2-one 97%
1-Methylimidazolidin-2-one

Precaución: for Research Use Only

Formula: C₄H₈N₂O
MW: 100.121 g/mol
Boiling Pt: 289.8 °C (760 mm Hg)
Melting Pt: 122…123 °C
Flash Pt: 129.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00800601
CAS Number: 694-32-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR