1-Methylcyclopropanamine hydrochloride 97%

Supplier: Ambeed
Warning

A105931-100G A105931-10G A105931-250MG A105931-25G A105931-1G A105931-5G
103796-972EA 13 USD
103796-972 103822-720 76809-638 76809-640 76809-642 76809-644 77309-580 77309-582
1-Methylcyclopropanamine hydrochloride 97%
1-Methylcyclopropanamine hydrochloride

Precaución: for Research Use Only

Formula: C₄H₁₀ClN
MW: 107.5800 g/mol
Melting Pt: 203…207 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11109576
CAS Number: 88887-87-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR