1-Methylcyclobutanecarboxylic acid 97%

Supplier: Ambeed
Warning

A217977-100MG A217977-250MG A217977-1G A217977-5G
103818-676EA 17 USD
103818-676 103849-408 76814-816 76814-818 77088-056 77088-058
1-Methylcyclobutanecarboxylic acid 97%
1-Methylcyclobutanecarboxylic acid

Precaución: for Research Use Only

Formula: C₆H₁₀O₂
MW: 114.14 g/mol
Boiling Pt: 203.5 °C (760 mm Hg)
Flash Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD15527494
CAS Number: 32936-76-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR