1-Methylcyclobutan-1-amine hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: 1-Methylcyclobutanamine hydrochloride

A172944-100MG A172944-250MG
103806-766EA 26 USD
103806-766 103845-058 76998-274 76998-276
1-Methylcyclobutan-1-amine hydrochloride 97%
1-Methylcyclobutan-1-amine hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₁N·ClH
MW: 121.608 g/mol
Melting Pt: 241…244 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09997048
CAS Number: 174886-05-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR