1-Methylazetidine-3-carboxylic acid 97%

Supplier: Ambeed
Warning

A167371-250MG A167371-1G A167371-25G A167371-5G
103806-176EA 15 USD
103806-176 103844-436 76973-842 76973-844 77081-444 77081-446 77081-448
1-Methylazetidine-3-carboxylic acid 97%
1-Methylazetidine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₉NO₂
Flash Pt: 79.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11520621
CAS Number: 875629-26-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR