N-Methyl-ε-caprolactam 98%

Supplier: Ambeed
Warning

Synonyms: N-Methyl-2-azacycloheptanone, 1-Methylazepan-2-one

A216263-100G A216263-10G A216263-25G A216263-5G
103849-262EA 7 USD
103849-262 76721-828 76721-830 77087-834 77087-836
N-Methyl-ε-caprolactam 98%
N-Methyl-epsilon-caprolactam

Precaución: for Research Use Only

Formula: C₇H₁₃NO
MW: 127.19 g/mol
Boiling Pt: 247.8 °C (760 mm Hg)
Flash Pt: 103.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00003263
CAS Number: 2556-73-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR