1-Methyl-6-oxo-1,6-dihydropyridine-3-carboxamide 98%

Supplier: Ambeed
Warning

A130732-250MG A130732-1G A130732-100MG
103801-468EA 119 USD
103801-468 77009-912 77009-914 77298-428 77889-892
1-Methyl-6-oxo-1,6-dihydropyridine-3-carboxamide 98%
1-Methyl-6-oxo-1,6-dihydropyridine-3-carboxamide

Precaución: for Research Use Only

Formula: C₇H₈N₂O₂
Boiling Pt: 396.7 °C (760 mm Hg)
Flash Pt: 193.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00457283
CAS Number: 701-44-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR