1-Methyl-5-(trifluoromethyl)-1H-pyrazol-3-ol 97%

Supplier: Ambeed
Warning

Synonyms: 1-Methyl-5-(trifluoromethyl)-1H-pyrazol-3(2H)-one

A768612-1G A768612-5G A768612-100MG A768612-250MG
103840-924EA 119 USD
103840-924 76845-166 76845-168 77243-996 77243-998
1-Methyl-5-(trifluoromethyl)-1H-pyrazol-3-ol 97%
1-Methyl-5-(trifluoromethyl)-1H-pyrazol-3-ol

Precaución: for Research Use Only

Formula: C₅H₅F₃N₂O
MW: 166.1 g/mol
Melting Pt: 129…134 °C
Storage Temperature: Ambient
MDL Number: MFCD00085094
CAS Number: 119022-51-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR