1-Methyl-4-piperidinemethanol 97%

Supplier: Ambeed
Warning

A229231-1G A229231-25G A229231-5G A229231-100G A229231-10G A229231-500G
103819-456EA 5 USD
103819-456 103850-356 77089-180 77089-182 77089-184 77112-920 77112-922 77112-924
1-Methyl-4-piperidinemethanol 97%
1-Methyl-4-piperidinemethanol

Precaución: for Research Use Only

Formula: C₇H₁₅NO
Boiling Pt: 189 °C (760 mm Hg)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD03411903
CAS Number: 20691-89-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR