1-Methyl-4-nitroimidazole 95%

Supplier: Ambeed
Warning

A289177-1G A289177-5G A289177-250MG
103813-818EA 50 USD
103813-818 103834-958 76837-530 76837-532 77668-282
1-Methyl-4-nitroimidazole 95%
1-Methyl-4-nitroimidazole

Precaución: for Research Use Only

Formula: C₄H₅N₃O₂
Boiling Pt: 328.9 °C (760 mm Hg)
Flash Pt: 152.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00159683
CAS Number: 3034-41-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR