2-Methyl-5-nitrobenzotrifluoride 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-Methyl-4-nitro-2-(trifluoromethyl)benzene
A173382-1G
A173382-25G
A173382-100G
A173382-10G
103806-814EA
9
USD
103806-814
103845-110
77082-354
77082-356
77236-326
77746-141
2-Methyl-5-nitrobenzotrifluoride 97%
2-Methyl-5-nitrobenzotrifluoride
Precaución: for Research Use Only
Formula:
C₈H₆F₃NO₂ MW: 205.14 g/mol Boiling Pt: 220.5 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD01631684 CAS Number: 89976-12-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...