1-Methyl-4-nitro-1H-pyrazole 98%

Supplier: Ambeed
Warning

Synonyms: 1-Methyl-4-nitropyrazole

A129692-100G A129692-10G A129692-1G A129692-25G A129692-5G
103801-312EA 5 USD
103801-312 76996-288 76996-290 77298-360 77298-362 77313-700
1-Methyl-4-nitro-1H-pyrazole 98%
1-Methyl-4-nitro-1H-pyrazole

Precaución: for Research Use Only

Formula: C₄H₅N₃O₂
MW: 127.10 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00464261
CAS Number: 3994-50-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR