1-Methyl-1H-indole-4-carboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: 1-Methyl-4-indolecarboxylic Acid

A316702-1G A316702-100MG A316702-250MG
103825-112EA 59 USD
103825-112 103836-040 76838-876 77032-708 77032-710
1-Methyl-1H-indole-4-carboxylic acid 95%
1-Methyl-1H-indole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₉NO₂
MW: 175.19 g/mol
Boiling Pt: 389 °C (760 mm Hg)
Melting Pt: 243…245 °C (760 mm Hg)
Flash Pt: 189 °C
Storage Temperature: Ambient
MDL Number: MFCD08435874
CAS Number: 90924-06-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR