1-Methyl-4,5,6,7-tetrahydro-1H-indazole-3-carboxylic acid 97%

Supplier: Ambeed
Warning

A252927-100MG A252927-250MG
103812-062EA 182 USD
103812-062 103851-910 77014-034 77014-036
1-Methyl-4,5,6,7-tetrahydro-1H-indazole-3-carboxylic acid 97%
1-Methyl-4,5,6,7-tetrahydro-1H-indazole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₁₂N₂O₂
MW: 180.2 g/mol
Boiling Pt: 378.2±42.0 °C (760 mm Hg)
Melting Pt: 207.5…208.5 °C
Flash Pt: 182.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09901219
CAS Number: 32286-99-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR