α,α,α-Trifluoro-p-xylene 96%

Supplier: Ambeed
Danger

Synonyms: 4-(Trifluoromethyl)toulene, 1-Methyl-4-(trifluoromethyl)benzene, 4-Methylbenzotrifluoride

A115861-25G A115861-1G A115861-5G
103798-638EA 9 USD
103798-638 103824-500 77136-264 77297-402 77297-404
α,α,α-Trifluoro-p-xylene 96%
α,α,α-Trifluoro-p-xylene

Precaución: for Research Use Only

Formula: C₈H₇F₃
MW: 160.14 g/mol
Boiling Pt: 129 °C (760 mm Hg)
Flash Pt: 27 °C
Storage Temperature: Ambient
MDL Number: MFCD00075476
CAS Number: 6140-17-6
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR