1-Methyl-4-piperidin-4-ylpiperazine 98%

Supplier: Ambeed
Danger

Synonyms: 1-Methyl-4-(piperidin-4-yl)piperazine

A514097-10G A514097-500G A514097-1G A514097-25G A514097-5G
103828-436EA 5 USD
103828-436 103839-540 76805-092 76805-094 76843-522 76843-524 76843-526
1-Methyl-4-piperidin-4-ylpiperazine 98%
1-Methyl-4-piperidin-4-ylpiperazine

Precaución: for Research Use Only

Formula: C₁₀H₂₁N₃
MW: 183.3 g/mol
Boiling Pt: 260.9 °C (760 mm Hg)
Flash Pt: 113.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03274729
CAS Number: 53617-36-0
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR