1-Methyl-4-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]piperazine 97%

Supplier: Ambeed
Warning

Synonyms: 2-(4-Methylpiperazin-1-yl)pyridine-5-boronic acid pinacol ester, [6-(4-Methylpiperazin-1-yl)pyridin-3-yl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-Methyl-4-(5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl)piperazine, 6-(4-Methylpiperazin-1-yl)pyridine-3-boronic acid Pinacol ester

A554188-100MG A554188-250MG A554188-1G A554188-25G A554188-5G
103828-706EA 13 USD
103828-706 103839-838 76830-572 76830-574 76843-922 76843-924 76843-926
1-Methyl-4-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]piperazine 97%
1-Methyl-4-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]piperazine

Precaución: for Research Use Only

Formula: C₁₆H₂₆BN₃O₂
MW: 303.21 g/mol
Boiling Pt: 435.2±45.0 °C (760 mm Hg)
Flash Pt: 217 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07437995
CAS Number: 918524-63-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR