1-Methyl-1H-pyrazole-4-boronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 1-Methylpyrazole-4-boronic acid pinacol ester, 1-Methyl-4-pyrazole boronic acid pinacol ester, 1-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

A909767-5G A909767-100G A909767-10G A909767-500G A909767-1G A909767-25G
103841-368EA 7 USD
103841-368 76845-644 77244-756 77244-758 77244-760 77308-502 77308-504
1-Methyl-1H-pyrazole-4-boronic acid pinacol ester 98%
1-Methyl-1H-pyrazole-4-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₀H₁₇BN₂O₂
Boiling Pt: 308.3 °C (760 mm Hg)
Melting Pt: 59…64 °C
Flash Pt: 140.3 °C
Storage Temperature: Ambient
MDL Number: MFCD03789259
CAS Number: 761446-44-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR