1-Methyl-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 1-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2,3,6-tetrahydropyridine

A292351-1G A292351-25G A292351-5G A292351-100G A292351-10G A292351-250MG
103813-936EA 7 USD
103813-936 103835-076 76837-672 76837-674 76837-676 76875-498 76875-500 76875-502
1-Methyl-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester 98%
1-Methyl-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₂₂BNO₂
MW: 223.12 g/mol
Boiling Pt: 252.5 °C (760 mm Hg)
Flash Pt: 109.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11506069
CAS Number: 454482-11-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR