2-Methyl-5-propyl-2H-pyrazole-3-carboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: 1-Methyl-3-propyl-1H-pyrazole-5-carboxylic acid

A115646-10G A115646-250MG A115646-1G A115646-5G
103798-600EA 67 USD
103798-600 103824-458 76995-506 76995-508 77311-276 77311-278
2-Methyl-5-propyl-2H-pyrazole-3-carboxylic acid 95%
2-Methyl-5-propyl-2H-pyrazole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₁₂N₂O₂
MW: 168.2 g/mol
Boiling Pt: 330.3 °C (760 mm Hg)
Melting Pt: 145…147 °C
Flash Pt: 153.5 °C
Storage Temperature: Ambient
MDL Number: MFCD02169703
CAS Number: 139755-99-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR